missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(4-nitrophenyl)amine, TCI America™

Manufacturer:  TCI America T233325G

 View more versions of this product

Catalog No. T233325G



Specifications

Specifications

Tris(4-nitrophenyl)amine
280°C
20440-93-1
MFCD00024636
C1=CC(=CC=C1N(C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-])[N+](=O)[O-]
380.316
380.32
≥96.0% (HPLC)
Undesignated
25g
C18H12N4O6
LSNJBIDKQIRWRQ-UHFFFAOYSA-N
4-nitro-N,N-bis(4-nitrophenyl)aniline
88538
Crystal-Powder at 20°C

Chemical Identifiers

20440-93-1
380.316
LSNJBIDKQIRWRQ-UHFFFAOYSA-N
4-nitro-N,N-bis(4-nitrophenyl)aniline
C18H12N4O6
MFCD00024636
88538
C1=CC(=CC=C1N(C2=CC=C(C=C2)[N+](=O)[O-])C3=CC=C(C=C3)[N+](=O)[O-])[N+](=O)[O-]
Safety and Handling

Safety and Handling

TSCA : Yes

Documents