Please login to your online account to display your discounted pricing

Tris(4-iodophenyl)amine 98%, ACROS Organics™

$76.64 - $76.64

Chemical Identifiers

Chemical Name or Material Tris(4-iodophenyl)amine
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C18H12I3N
Linear Formula N(C6H4I)3
Formula Weight 623
MDL Number MFCD01321198
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391060010

  SDS
thermo fisher scientific
AC391060010
Glass bottle 1g Each for $76.64

AC391060050

  SDS
thermo fisher scientific
AC391060050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (HPLC)
Formula Weight 623
Physical Form Liquid
Quantity 1g
Color Clear colorless
Melting Point 186° to 190°C
Linear Formula N(C6H4I)3
Chemical Name or Material Tris(4-iodophenyl)amine
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS 4181-20-8
MDL Number MFCD01321198
Packaging Glass bottle
Molecular Formula C18H12I3N
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.