Learn More
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride, Thermo Scientific Chemicals
Redox indicator, oxygen sensor
Supplier: Thermo Scientific Chemicals 04412302
Description
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride is used as a probe for luminescent detection and quantitation of oxygen. It is also used for fiber optic sensors. Further, it is used in studies on oxygen in skin and skin tumors as well as measurement of oxygen flux through the skin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tris (4,7-diphenyl-1,10-phenanthroline)ruthenium (II) dichloride | |
C72H48Cl2N6Ru | |
0.5 g | |
Soluble in alcohol and dimethylformamide. Insoluble in water. | |
C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.Cl[Ru]Cl | |
1169.188 | |
1169.2 |
36309-88-3 | |
MFCD03095387 | |
bathophenanthroline ruthenium chloride, ru ddp, tris-bathophenanthroline ruthenium ii chloride, dichlororuthenium-4,7-diphenyl-1,10-phenanthroline 1/3, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride, tris 4,7-diphenyl-1,10-phenanthroline ; dichlororuthenium, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, for fluorescence | |
SKZWFYFFTOHWQP-UHFFFAOYSA-L | |
dichlororuthenium;4,7-diphenyl-1,10-phenanthroline | |
15787787 | |
Powder |
Chemical Identifiers
36309-88-3 | |
1169.188 | |
SKZWFYFFTOHWQP-UHFFFAOYSA-L | |
15787787 | |
C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5.Cl[Ru]Cl |
C72H48Cl2N6Ru | |
MFCD03095387 | |
bathophenanthroline ruthenium chloride, ru ddp, tris-bathophenanthroline ruthenium ii chloride, dichlororuthenium-4,7-diphenyl-1,10-phenanthroline 1/3, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride, tris 4,7-diphenyl-1,10-phenanthroline ; dichlororuthenium, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, tris 4,7-diphenyl-1,10-phenanthroline ruthenium ii dichloride complex, for fluorescence | |
dichlororuthenium;4,7-diphenyl-1,10-phenanthroline |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only