Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(3-fluorophenyl)phosphine, 97%

$132.28 - $587.64

Chemical Identifiers

Chemical Name or Material Tris(3-fluorophenyl)phosphine
CAS 23039-94-3
Molecular Formula C18H12F3P
Formula Weight 316.27
MDL Number MFCD00015722
Beilstein 2946854
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1511703

  SDS
alfa aesar™
A1511703
1g Each for $132.28

AAA1511706

  SDS
alfa aesar™
A1511706
5g Each for $587.64
Description & Specifications

Specifications

Formula Weight 316.27
Chemical Name or Material Tris(3-fluorophenyl)phosphine
Assay 97%
Molecular Formula C18H12F3P
MDL Number MFCD00015722
CAS 23039-94-3
Beilstein 2946854
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 60° to 63°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.