missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(3-fluorophenyl)phosphine, 97%

Manufacturer:  Alfa Aesar™ A1511703

 View more versions of this product

Catalog No. AAA1511703



Specifications

Specifications

Tris(3-fluorophenyl)phosphine
1g
C18H12F3P
2946854
CUTRINLXFPIWQB-UHFFFAOYSA-N
tris(3-fluorophenyl)phosphane
140988
97%
60°C to 63°C
23039-94-3
MFCD00015722
tris 3-fluorophenyl phosphine, tris 3-fluorophenyl phosphane, tri 3-fluorophenyl phosphine, phosphine,tris 3-fluorophenyl, acmc-1cqwl, tri m-fluorophenyl phosphine, tri-3-fluorophenyl-phosphine
C1=CC(=CC(=C1)P(C2=CC=CC(=C2)F)C3=CC=CC(=C3)F)F
316.263
316.27

Chemical Identifiers

23039-94-3
316.263
CUTRINLXFPIWQB-UHFFFAOYSA-N
140988
C1=CC(=CC(=C1)P(C2=CC=CC(=C2)F)C3=CC=CC(=C3)F)F
C18H12F3P
MFCD00015722
tris 3-fluorophenyl phosphine, tris 3-fluorophenyl phosphane, tri 3-fluorophenyl phosphine, phosphine,tris 3-fluorophenyl, acmc-1cqwl, tri m-fluorophenyl phosphine, tri-3-fluorophenyl-phosphine
tris(3-fluorophenyl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents