missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris(3,5-dimethyl-1-pyrazolyl)methane 98.0+%, TCI America™
Supplier: TCI America T30251G
Specifications
Tris(3,5-dimethyl-1-pyrazolyl)methane | |
152°C | |
C16H22N6 | |
1 g | |
CC1=NN(C(N2N=C(C)C=C2C)N2N=C(C)C=C2C)C(C)=C1 | |
298.39 | |
298.39 | |
Crystalline Powder |
28791-97-1 | |
White-Yellow | |
MFCD00958749 | |
COKUMKYWINNWLV-UHFFFAOYSA-N | |
1-[bis(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-3,5-dimethyl-1H-pyrazole | |
4359225 | |
≥98.0% (GC) |
Chemical Identifiers
28791-97-1 | |
298.39 | |
COKUMKYWINNWLV-UHFFFAOYSA-N | |
1-[bis(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-3,5-dimethyl-1H-pyrazole |
C16H22N6 | |
MFCD00958749 | |
4359225 | |
CC1=NN(C(N2N=C(C)C=C2C)N2N=C(C)C=C2C)C(C)=C1 |
Safety and Handling
TSCA : No