Please login to your online account to display your discounted pricing

Tris(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)phenyltin, 95%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  333810010

 View more versions of this product

Catalog No. AC333810010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94.0%
Physical Form Slightly Viscous Liquid
Formula Weight 1237.11
Quantity 1g
Color Clear colorless
Chemical Name or Material Tris(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)phenyltin
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C30H17F39Sn
MDL Number MFCD01863239
CAS 175354-30-0
Refractive Index 1.3690 to 1.3710 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tris(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)phenyltin
CAS 175354-30-0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C30H17F39Sn
Formula Weight 1237.11
MDL Number MFCD01863239
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.

SDS & Safety Documents