Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2-thienyl)phosphine, 98+%

$67.81 - $967.34

Chemical Identifiers

Chemical Name or Material Tris(2-thienyl)phosphine
CAS 24171-89-9
Molecular Formula C12H9PS3
Formula Weight 280.37
MDL Number MFCD00015471
Beilstein 203736
Synonym Tri(2-thienyl)phosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1337903

  SDS
alfa aesar™
A1337903
1g Each for $67.81

AAA1337906

  SDS
alfa aesar™
A1337906
5g Each for $233.55

AAA1337914

  SDS
alfa aesar™
A1337914
25g Each for $967.34
Description & Specifications

Specifications

Formula Weight 280.37
Chemical Name or Material Tris(2-thienyl)phosphine
Assay ≥98%
Molecular Formula C12H9PS3
MDL Number MFCD00015471
CAS 24171-89-9
Beilstein 203736
Synonym Tri(2-thienyl)phosphine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 28° to 30°C
Odor Recognizable
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool, dry place in tightly closed container. Store under dry inert gas.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.