missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(2-pyridylmethyl)amine, TCI America™

Manufacturer:  TCI America T26711G

 View more versions of this product

Catalog No. T26711G



Specifications

Specifications

Tris(2-pyridylmethyl)amine
86°C
16858-01-8
VGUWFGWZSVLROP-UHFFFAOYSA-N
1-pyridin-2-yl-N,N-bis(pyridin-2-ylmethyl)methanamine
379259
Crystal-Powder at 20°C
White-Yellow
1g
C18H18N4
C1=CC=NC(=C1)CN(CC2=CC=CC=N2)CC3=CC=CC=N3
290.37
290.37
≥98.0% (HPLC)

Chemical Identifiers

16858-01-8
290.37
379259
C1=CC=NC(=C1)CN(CC2=CC=CC=N2)CC3=CC=CC=N3
C18H18N4
VGUWFGWZSVLROP-UHFFFAOYSA-N
1-pyridin-2-yl-N,N-bis(pyridin-2-ylmethyl)methanamine
Safety and Handling

Safety and Handling

TSCA : No

Documents