Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2-phenylpyridine)iridium(III)

As a green light emitting material

$226.01 - $226.01

Chemical Identifiers

Chemical Name or Material Tris (2-phenylpyridine)iridium (III)
CAS 693794-98-8
Molecular Formula C33H24IrN3
Formula Weight 654.82
MDL Number MFCD04039766
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4428277

  SDS
alfa aesar™
4428277
0.1g Each for $226.01
Description & Specifications

Specifications

Chemical Name or Material Tris (2-phenylpyridine)iridium (III)
CAS 693794-98-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
Molecular Formula C33H24IrN3
Formula Weight 654.82
MDL Number MFCD04039766
Color Yellow
Melting Point 326° to 328°C
Quantity 0.1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.