missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2-methoxyethoxy)vinylsilane, 98%

Manufacturer:  Alfa Aesar™ H60717AP

 View more versions of this product

Catalog No. AAH60717AP



Specifications

Specifications

Tris(2-methoxyethoxy)vinylsilane
C11H24O6Si
1795316
WOXXJEVNDJOOLV-UHFFFAOYSA-N
ethenyl-tris(2-methoxyethoxy)silane
14025
Liquid
1.034g/mL
136°C (5.5mmHg)
500mL
1067-53-4
MFCD00008500
2,5,7,10-tetraoxa-6-silaundecane, 6-ethenyl-6-2-methoxyethoxy
COCCO[Si](C=C)(OCCOC)OCCOC
280.392
280.39
98%
Moisture Sensitive
Brown to Colorless

Chemical Identifiers

1067-53-4
280.392
WOXXJEVNDJOOLV-UHFFFAOYSA-N
14025
COCCO[Si](C=C)(OCCOC)OCCOC
C11H24O6Si
MFCD00008500
2,5,7,10-tetraoxa-6-silaundecane, 6-ethenyl-6-2-methoxyethoxy
ethenyl-tris(2-methoxyethoxy)silane
Safety and Handling

Safety and Handling

GHS H Statement
H361-H312-H319
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement
P280-P281-P305+P351+P338-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VV6826000

TSCA : TSCA

Documents