Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2-methoxyethoxy)vinylsilane, 98%

Chemical Identifiers

Chemical Name or Material Tris(2-methoxyethoxy)vinylsilane
CAS 1067-53-4
Molecular Formula C11H24O6Si
Formula Weight 280.39
MDL Number MFCD00008500
Beilstein 1795316
Synonym Vinyltris(2-methoxyethoxy)silane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH60717AP

  SDS
alfa aesar™
H60717AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.39
Molecular Formula C11H24O6Si
Chemical Name or Material Tris(2-methoxyethoxy)vinylsilane
TSCA TSCA
Physical Form Liquid
Assay 98%
Sensitivity Moisture Sensitive
MDL Number MFCD00008500
CAS 1067-53-4
Beilstein 1795316
Synonym Vinyltris(2-methoxyethoxy)silane
Boiling Point 136°C (5.5mmHg)
Color Colorless to pale brown
Quantity 500mL
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H361-H312-H319
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P281-P305+P351+P338-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.034g/mL
RTECS Number VV6826000
Safety and Handling

Warning

GHS P Statement
P280-P281-P305+P351+P338-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H361-H312-H319
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Causes serious eye irritation.