Please login to your online account to display your discounted pricing

Tris (2-cyanoethyl)phosphine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material Tris (2-cyanoethyl)phosphine
Assay Percent Range 95%
Molecular Formula C9H12N3P
Formula Weight 193.19
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC449710010

  SDS
thermo fisher scientific
AC449710010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC449710500

  SDS
thermo fisher scientific
AC449710500
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC449710100

  SDS
thermo fisher scientific
AC449710100
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tris (2-cyanoethyl)phosphine
Quantity 1g
CAS 4023-53-4
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C9H12N3P
Formula Weight 193.19