missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris (2-cyanoethyl)phosphine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC449710500
Specifications
Tris (2-cyanoethyl)phosphine | |
235.0°C (0.9 mmHg) | |
50g | |
94% min. (Iodimetry) | |
3,3',3-phosphinetriyltripropanenitrile, 3-bis 2-cyanoethyl phosphanyl propanenitrile, acmc-20ajc3, p ch2ch2cn 3, propanenitrile, 3,3',3-phosphinidynetris, propanenitrile,3',3-phosphinidynetris, tris 2-cyanoethyl phosphine, tris 2-cyanoethyl phosphine 10g, tris-2-cyanoethyl phosphine, tris-2-cyanoethyl-phosphine | |
CHZAMJVESILJGH-UHFFFAOYSA-N | |
3-[bis(2-cyanoethyl)phosphanyl]propanenitrile | |
77639 | |
95% |
Glass bottle | |
97.0°C to 99.0°C | |
4023-53-4 | |
C9H12N3P | |
Solubility in water: slightly soluble | |
C(CP(CCC#N)CCC#N)C#N | |
193.19 | |
193.19 |
Chemical Identifiers
4023-53-4 | |
193.19 | |
3,3',3-phosphinetriyltripropanenitrile, 3-bis 2-cyanoethyl phosphanyl propanenitrile, acmc-20ajc3, p ch2ch2cn 3, propanenitrile, 3,3',3-phosphinidynetris, propanenitrile,3',3-phosphinidynetris, tris 2-cyanoethyl phosphine, tris 2-cyanoethyl phosphine 10g, tris-2-cyanoethyl phosphine, tris-2-cyanoethyl-phosphine | |
3-[bis(2-cyanoethyl)phosphanyl]propanenitrile |
C9H12N3P | |
CHZAMJVESILJGH-UHFFFAOYSA-N | |
77639 | |
C(CP(CCC#N)CCC#N)C#N |
Safety and Handling
GHS H Statement
May cause cancer.
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Obtain special instructions before use.
Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Danger
EINECSNumber : 223-687-