Please login to your online account to display your discounted pricing

Tris(2-chloroethyl) phosphate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  152810250

 View more versions of this product

Catalog No. AC152810250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (GC)
Physical Form Liquid
Formula Weight 285.48
Quantity 25g
Color Clear colorless
EINECS Number 204-118-5
TSCA TSCA
Density 1.3900g/mL
RTECS Number KK2450000
Beilstein 01, II, 337
Refractive Index 1.4710 to 1.4740 (20°C, 589nm)
Chemical Name or Material Tris(2-chloroethyl) phosphate
Name Note 97%
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H12Cl3O4P
Linear Formula (ClCH2CH2O)3P(O)
Packaging Glass bottle
MDL Number MFCD00000967
CAS 115-96-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Suspected of causing cancer.
May damage fertility.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tris(2-chloroethyl) phosphate
Name Note 97%
CAS 115-96-8
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C6H12Cl3O4P
Linear Formula (ClCH2CH2O)3P(O)
Formula Weight 285.48
MDL Number MFCD00000967
Accessories

Danger

GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Suspected of causing cancer.
May damage fertility.
Harmful if swallowed.

SDS & Safety Documents