Please login to your online account to display your discounted pricing

Tris(2-chloroethyl) phosphate, 97%, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  152810010

 View more versions of this product

Catalog No. AC152810010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 285.48
Assay ≥96.0% (GC)
Quantity 1kg
Color Clear colorless
Beilstein 01, II, 337
RTECS Number KK2450000
TSCA TSCA
EINECS Number 204-118-5
Density 1.3900g/mL
Infrared Spectrum Authentic
Refractive Index 1.4710 to 1.4740 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Suspected of causing cancer.
May damage fertility.
Harmful if swallowed.
Name Note 97%
Chemical Name or Material Tris(2-chloroethyl) phosphate
Linear Formula (ClCH2CH2O)3P(O)
CAS 115-96-8
Molecular Formula C6H12Cl3O4P
Assay Percent Range 97%
MDL Number MFCD00000967

Specifications

Chemical Name or Material Tris(2-chloroethyl) phosphate
Name Note 97%
CAS 115-96-8
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C6H12Cl3O4P
Linear Formula (ClCH2CH2O)3P(O)
Formula Weight 285.48
MDL Number MFCD00000967
Accessories

Danger

GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Suspected of causing cancer.
May damage fertility.
Harmful if swallowed.

SDS & Safety Documents