Please login to your online account to display your discounted pricing

Tris(2-carboxyethyl)phosphine hydrochloride, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  363830010

 View more versions of this product

Catalog No. AC363830010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 286.65
Quantity 1g
Color White
Merck Index 15, 9222
Beilstein 04, IV, 3441
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00145469
Assay Percent Range 98%
CAS 51805-45-9
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
Molecular Formula C9H15O6P·HCl

Specifications

Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
CAS 51805-45-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H15O6P·HCl
Formula Weight 286.65
MDL Number MFCD00145469
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents