Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2-carboxyethyl)phosphine hydrochloride, 98%

Reducing agent, specific for disulfides

$136.08 - $531.05

Chemical Identifiers

Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
CAS 51805-45-9
Linear Formula (HO2CCH2CH2)3P·HCl
Formula Weight 286.65
MDL Number MFCD00145469
UN Number UN3261
Merck Index 14,9085
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4058704

 
alfa aesar™
4058704
2g Each for $136.08

AA4058709

 
alfa aesar™
4058709
10g Each for $531.05

AA4058702

 
alfa aesar™
4058702
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 286.65
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
Assay 98%
Linear Formula (HO2CCH2CH2)3P·HCl
MDL Number MFCD00145469
CAS 51805-45-9
UN Number UN3261
Merck Index 14,9085
Solubility Information Soluble in water
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Color White
Melting Point 177° to 178°C
Quantity 2g
Sensitivity Moisture Sensitive
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.