Learn More
Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H51864AA
Description
Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.
Solubility
Miscible with water and methanol.
Notes
Air sensitive. Store in cool place. Incompatible with strong oxidizing agents
Specifications
Tris(2-carboxyethyl)phosphine hydrochloride | |
C9H12O6P | |
MFCD00145469 | |
UN3265 | |
Miscible with water and methanol. | |
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O | |
247.16 | |
286.65 | |
0.5M soln. in water |
51805-45-9 | |
(HO2CCH2CH2)3P·HCl | |
5 mL | |
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride | |
PZBFGYYEXUXCOF-UHFFFAOYSA-K | |
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride | |
2734570 | |
95% |
Chemical Identifiers
51805-45-9 | |
247.16 | |
PZBFGYYEXUXCOF-UHFFFAOYSA-K | |
2734570 | |
[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O |
C9H12O6P | |
MFCD00145469 | |
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride | |
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only