Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2-aminoethyl)amine, 97%

$86.05 - $237.05

Chemical Identifiers

Chemical Name or Material Tris(2-aminoethyl)amine
CAS 4097-89-6
Molecular Formula C6H18N4
Linear Formula N(CH2CH2NH2)3
Formula Weight 146.24
MDL Number MFCD00008177
UN Number UN2922
Beilstein 1739626
Synonym 2,2',2''-Triaminotriethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2178914

  SDS
alfa aesar™
B2178914*
25g Each for $86.05

AAB2178922

  SDS
alfa aesar™
B2178922
100g Each for $237.05

AAB2178936

  SDS
alfa aesar™
B2178936
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.24
Chemical Name or Material Tris(2-aminoethyl)amine
TSCA TSCA
Assay 97%
Molecular Formula C6H18N4
Linear Formula N(CH2CH2NH2)3
MDL Number MFCD00008177
CAS 4097-89-6
UN Number UN2922
Beilstein 1739626
Synonym 2,2',2''-Triaminotriethylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H310-H314-H318
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
pH 11.9
Boiling Point 102°C (0.05mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Quantity 25g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECS Number KH8587082

Applications
Tris(2-aminoethyl)amine is a tetradentate chelating ligand and forms stable complexes with transition metals. It is also used as a carbon dioxide absorbent. Further, it acts as a reagent for cleavage of the fluorenylmethyloxycarbonyl (Fmoc) group in peptide synthesis. It reacts with aryl isocyanates and isothiocyanates to give tris-urea and -thiourea derivatives.

Solubility
Miscible with water.

Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with strong acids and strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H310-H314-H318
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.