Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,6-dimethoxyphenyl)phosphine, 98%

$82.35 - $82.35

Chemical Identifiers

Chemical Name or Material Tris(2,6-dimethoxyphenyl)phosphine
CAS 85417-41-0
Molecular Formula C24H27O6P
Formula Weight 442.45
MDL Number MFCD00192558
Synonym TDMPP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1516206

 
alfa aesar™
L1516206
5g Each for $82.35
Description & Specifications

Specifications

Formula Weight 442.45
Chemical Name or Material Tris(2,6-dimethoxyphenyl)phosphine
Molecular Formula C24H27O6P
MDL Number MFCD00192558
CAS 85417-41-0
Synonym TDMPP
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Melting Point 149° to 156°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.