missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(2,4-pentanedionato)manganese(III) 98.0+%, TCI America™

Supplier:  TCI America M004325G

 View more versions of this product

Catalog No. M004325G



Description

Description

Specifications

Specifications

Tris(2,4-pentanedionato)manganese(III)
161°C
C15H21MnO6
25 g
HYZQBNDRDQEWAN-LNTINUHCSA-K
manganese(3+) tris((2Z)-4-oxopent-2-en-2-olate)
122130696
≥98.0% (T)
14284-89-0
Black
MFCD00000023
mn iii acetylacetonate, tris acetylacetonato manganese iii
[Mn+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
352.27
352.27
Crystalline Powder

Chemical Identifiers

14284-89-0
352.27
HYZQBNDRDQEWAN-LNTINUHCSA-K
122130696
[Mn+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
C15H21MnO6
MFCD00000023
mn iii acetylacetonate, tris acetylacetonato manganese iii
manganese(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2142

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions