missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(2,4-pentanedionato)manganese(III), TCI America™

Manufacturer:  TCI America M0043100G

 View more versions of this product

Catalog No. M0043100G



Specifications

Specifications

Tris(2,4-pentanedionato)manganese(III)
161°C
14284-89-0
MFCD00000023
BCOBLSSFEZDWKH-BJILWQEISA-M
manganese(3+);(E)-4-oxopent-2-en-2-olate
122130696
Crystal-Powder at 20°C
Black
100g
C5H7MnO2+2
mn iii acetylacetonate, tris acetylacetonato manganese iii
CC(=CC(=O)C)[O-].[Mn+3]
154.047
352.27
≥98.0% (T)

Chemical Identifiers

14284-89-0
154.047
BCOBLSSFEZDWKH-BJILWQEISA-M
122130696
CC(=CC(=O)C)[O-].[Mn+3]
C5H7MnO2+2
MFCD00000023
mn iii acetylacetonate, tris acetylacetonato manganese iii
manganese(3+);(E)-4-oxopent-2-en-2-olate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2142

TSCA : Yes

Documents