missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris(2,2'-bipyridyl)ruthenium(II) Chloride Hexahydrate 98.0+%, TCI America™
Supplier: TCI America T16551G
Specifications
Tris(2,2′-bipyridyl)ruthenium(II) Chloride Hexahydrate | |
Red-Yellow | |
MFCD00149670 | |
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1 | |
748.62 | |
640.53 | |
Crystalline Powder |
50525-27-4 | |
C30H36Cl2N6O6Ru | |
1 g | |
WHELTKFSBJNBMQ-UHFFFAOYSA-L | |
λ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride | |
131664161 | |
≥98.0% (T) |
Chemical Identifiers
50525-27-4 | |
748.62 | |
WHELTKFSBJNBMQ-UHFFFAOYSA-L | |
131664161 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1 |
C30H36Cl2N6O6Ru | |
MFCD00149670 | |
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate | |
λ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride |
Safety and Handling
RTECSNumber : VM2730000
TSCA : No