missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 208750010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tris(2, 2'-bipyridyl)ruthenium(II) chloride, hexahydrate | |
>300.0°C | |
Authentic | |
Glass bottle | |
MFCD00149670 | |
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate | |
WHELTKFSBJNBMQ-UHFFFAOYSA-L | |
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate | |
131664161 | |
98% |
50525-27-4 | |
97.5% min. | |
98% | |
C30H36Cl2N6O6Ru | |
1 g | |
Solubility in water: soluble. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1 | |
748.62 | |
748.63 | |
Crystalline Powder or Crystals |
Chemical Identifiers
50525-27-4 | |
748.62 | |
WHELTKFSBJNBMQ-UHFFFAOYSA-L | |
131664161 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1 |
C30H36Cl2N6O6Ru | |
MFCD00149670 | |
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate | |
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate |
RUO – Research Use Only