Learn More
Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III), recrystallized, 99.9+% (metals basis)
Manufacturer: Alfa Aesar™ 4400303
Specifications
Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium (III) | |
170°C to 173°C | |
Recrystallized | |
(metals basis) | |
MFCD00015713 | |
PPRRRPCEDUWEHL-LWTKGLMZSA-K | |
(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;yttrium(3+) | |
50908983 | |
Crystalline |
95°C (0.05mmHg) | |
1g | |
15632-39-0 | |
C33H57O6Y | |
y tmhd 3 | |
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Y+3] | |
638.719 | |
638.7 | |
≥99.9% |
Chemical Identifiers
15632-39-0 | |
638.719 | |
PPRRRPCEDUWEHL-LWTKGLMZSA-K | |
50908983 | |
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Y+3] |
C33H57O6Y | |
MFCD00015713 | |
y tmhd 3 | |
(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;yttrium(3+) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning