Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III), recrystallized, 99.9+% (metals basis)

$70.35 - $1,023.62

Chemical Identifiers

Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium (III)
CAS 15632-39-0
Name Note Recrystallized
Molecular Formula Y[(CH3)3CCOCH=COC(CH3)3]3
Formula Weight 638.7
MDL Number MFCD00015713
Synonym Yttrium(III)-DPM; Y(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4400303

  SDS
alfa aesar™
4400303
1g Each for $70.35

AA4400306

  SDS
alfa aesar™
4400306
5g Each for $278.61

AA4400314

  SDS
alfa aesar™
4400314
25g Each for $1,023.62
Description & Specifications

Specifications

Formula Weight 638.7
Name Note Recrystallized
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium (III)
Physical Form Crystalline
Assay ≥99.9%
Assay Percent Notes (metals basis)
Molecular Formula Y[(CH3)3CCOCH=COC(CH3)3]3
MDL Number MFCD00015713
CAS 15632-39-0
Synonym Yttrium(III)-DPM; Y(TMHD)3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 170° to 173°C
Boiling Point 95°C (0.05mmHg)
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.