Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III), 98+%

$343.26 - $343.26

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III)
CAS 15632-39-0
Formula Weight 638.7
MDL Number MFCD00015713
Synonym Yttrium(III)-DPM; Y(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1448003

  SDS
alfa aesar™
1448003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1448006

  SDS
alfa aesar™
1448006
5g Each for $343.26
Description & Specifications

Specifications

Formula Weight 638.7
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III)
Physical Form Crystalline
Assay ≥98%
MDL Number MFCD00015713
CAS 15632-39-0
Synonym Yttrium(III)-DPM; Y(TMHD)3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 170° to 173°C
Boiling Point 95°C (0.05mmHg)
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.