missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III), 98+%

Manufacturer:  Alfa Aesar™ 1448003

 View more versions of this product

Catalog No. AA1448003



Specifications

Specifications

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III)
95°C (0.05mmHg)
1g
C33H57O6Y
y tmhd 3
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Y+3]
638.719
638.7
≥98%
Hygroscopic
170°C to 173°C
15632-39-0
MFCD00015713
PPRRRPCEDUWEHL-LWTKGLMZSA-K
(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;yttrium(3+)
50908983
Crystalline

Chemical Identifiers

15632-39-0
638.719
PPRRRPCEDUWEHL-LWTKGLMZSA-K
50908983
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Y+3]
C33H57O6Y
MFCD00015713
y tmhd 3
(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;yttrium(3+)
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents