missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), 99%

$91.91 - $91.91

Chemical Identifiers

CAS 15492-52-1
Formula Weight 722.86
MDL Number MFCD00000034
Synonym Ytterbium(III)-DPM; Yb(TMHD)3
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3958103 SDS Alfa Aesar™
3958103
1g Each for $91.91
AA3958106 SDS Alfa Aesar™
3958106
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

15492-52-1
MFCD00000034
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III)
722.86
Ytterbium(III)-DPM; Yb(TMHD)3

Specifications

15492-52-1
722.86
Ytterbium(III)-DPM; Yb(TMHD)3
1g
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III)
Crystalline
MFCD00000034
167° to 169°C
Hygroscopic
99%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA