Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)terbium(III), 99%

$100.96 - $474.64

Chemical Identifiers

Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)terbium (III)
CAS 15492-51-0
Molecular Formula C33H57O6Tb
Formula Weight 708.74
MDL Number MFCD00064752
Synonym Terbium(III)-DPM; Tb(TMHD3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4484203

  SDS
alfa aesar™
4484203
1g Each for $100.96

AA4484206

  SDS
alfa aesar™
4484206
5g Each for $474.64
Description & Specifications

Specifications

Formula Weight 708.74
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)terbium (III)
Physical Form Granular
Assay 99%
Molecular Formula C33H57O6Tb
MDL Number MFCD00064752
CAS 15492-51-0
Synonym Terbium(III)-DPM; Tb(TMHD3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 155° to 156°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.