missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)terbium(III), 99%

Manufacturer:  Alfa Aesar™ 4484203

 View more versions of this product

Catalog No. AA4484203



Specifications

Specifications

Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)terbium (III)
1g
C33H57O6Tb
tb tmhd 3, terbium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Tb+3]
708.738
708.74
99%
155°C to 156°C
15492-51-0
MFCD00064752
JSAHJYGCYOFNGW-LWTKGLMZSA-K
terbium(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
133109942
Granular

Chemical Identifiers

15492-51-0
708.738
JSAHJYGCYOFNGW-LWTKGLMZSA-K
133109942
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Tb+3]
C33H57O6Tb
MFCD00064752
tb tmhd 3, terbium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate
terbium(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents