missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)scandium(III), 95+%

Intermediate for scandium oxide ceramics

$102.99 - $315.55

Chemical Identifiers

CAS 15492-49-6
Molecular Formula C33H57O6Sc
Formula Weight 594.77
MDL Number MFCD00169875
Synonym Scandium(III)-DPM; Sc(TMHD)3
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)scandium (III)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4278702 SDS Alfa Aesar™
4278702
0.5g Each for $102.99
AA4278704 SDS Alfa Aesar™
4278704
2g Each for $315.55
Specifications Description & Specifications

Chemical Identifiers

15492-49-6
594.77
Scandium(III)-DPM; Sc(TMHD)3
C33H57O6Sc
MFCD00169875
Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)scandium (III)

Specifications

15492-49-6
C33H57O6Sc
MFCD00169875
Soluble in ethyl acetate
150° to 152°C
>110°C (230°F)
Hygroscopic
≥95%
Powder
594.77
Scandium(III)-DPM; Sc(TMHD)3
White
275°C (dec.)
0.5g
Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)scandium (III)

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning