Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ruthenium(III), 99%

$232.04 - $810.64

Chemical Identifiers

Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)ruthenium (III)
CAS 38625-54-6
Molecular Formula C33H57O6Ru
Formula Weight 650.88
MDL Number MFCD00269841
Synonym Ruthenium(III)-DPM; Ru(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4400602

  SDS
alfa aesar™
4400602
0.5g Each for $232.04

AA4400604

  SDS
alfa aesar™
4400604
2g Each for $810.64
Description & Specifications

Specifications

Formula Weight 650.88
Physical Form Powder
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym Ruthenium(III)-DPM; Ru(TMHD)3
Melting Point 200° to 203°C
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)ruthenium (III)
Boiling Point 250°C (dec.)
MDL Number MFCD00269841
Health Hazard 1 Warning
CAS 38625-54-6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C33H57O6Ru
Sensitivity Hygroscopic
Assay 99%
Quantity 0.5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.