missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ruthenium(III), 99%

Manufacturer:  Alfa Aesar™ 4400604

 View more versions of this product

Catalog No. AA4400604



Specifications

Specifications

Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)ruthenium (III)
250°C (decomposition)
2g
C33H60O6Ru
tris 2,2,6,6-tetramethyl-3,5-heptanedionato ruthenium iii
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Ru]
653.907
650.88
99%
Hygroscopic
200°C to 203°C
38625-54-6
MFCD00269841
TZPFVUZFVRIDGB-LWTKGLMZSA-N
(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;ruthenium
131675815
Powder

Chemical Identifiers

38625-54-6
653.907
TZPFVUZFVRIDGB-LWTKGLMZSA-N
131675815
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Ru]
C33H60O6Ru
MFCD00269841
tris 2,2,6,6-tetramethyl-3,5-heptanedionato ruthenium iii
(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;ruthenium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents