Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium(III), 99%

$46.80 - $201.65

Chemical Identifiers

Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium (III)
CAS 15492-48-5
Molecular Formula C33H57O6Pr
Formula Weight 690.72
MDL Number MFCD00000027
Synonym Praseodymium(III)-DPM; Pr(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1225903

 
alfa aesar™
A1225903
1g Each for $46.80

AAA1225906

 
alfa aesar™
A1225906
5g Each for $201.65
Description & Specifications

Specifications

Formula Weight 690.72
TSCA TSCA
Assay 99%
Melting Point 219° to 221°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium (III)
Molecular Formula C33H57O6Pr
MDL Number MFCD00000027
CAS 15492-48-5
Synonym Praseodymium(III)-DPM; Pr(TMHD)3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.