Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)neodymium(III)

$76.50 - $76.50

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)neodymium(III)
CAS 15492-47-4
Formula Weight 694.06g/mol
MDL Number MFCD00015708
Synonym Neodymium(III)-DPM; Nd(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3929803

 
alfa aesar™
39298-03
1g Each for $76.50
Description & Specifications

Specifications

Melting Point 209° to 212°C
Boiling Point 270°C (dec.)
Quantity 1g
Sensitivity Hygroscopic
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)neodymium(III)
CAS 15492-47-4
Physical Form Crystalline
Formula Weight 694.06g/mol
MDL Number MFCD00015708
Synonym Neodymium(III)-DPM; Nd(TMHD)3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.