Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III), 98%

$77.15 - $77.15

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)
CAS 14319-13-2
Formula Weight 688.72
MDL Number MFCD00010466
Synonym Lanthanum(III)-DPM; La(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3646503

  SDS
alfa aesar™
3646503
1g Each for $77.15

AA3646506

  SDS
alfa aesar™
3646506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 688.72
Synonym Lanthanum(III)-DPM; La(TMHD)3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
MDL Number MFCD00010466
CAS 14319-13-2
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)
Sensitivity Hygroscopic
Boiling Point 370°C (dec.)
Melting Point 240°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Physical Form Powder
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.