Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)holmium(III)

$108.18 - $326.97

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)holmium(III)
CAS 15522-73-3
Formula Weight 714.73
MDL Number MFCD00070457
Synonym Holmium(III)-DPM; Ho(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3353103

  SDS
alfa aesar™
3353103
1g Each for $108.18

AA3353106

  SDS
alfa aesar™
3353106
5g Each for $326.97
Description & Specifications

Specifications

Formula Weight 714.73
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 15522-73-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Melting Point 179° to 183°C
Synonym Holmium(III)-DPM; Ho(TMHD)3
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)holmium(III)
Health Hazard 1 Warning
Sensitivity Hygroscopic
Physical Form Crystalline
MDL Number MFCD00070457
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.