Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III)

$117.61 - $429.46

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III)
CAS 34228-15-4
Formula Weight 619.54
MDL Number MFCD00058705
Synonym Gallium(III)-DPM; Ga(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1449103

  SDS
alfa aesar™
1449103
1g Each for $117.61

AA1449106

  SDS
alfa aesar™
1449106
5g Each for $429.46
Description & Specifications

Specifications

Formula Weight 619.54
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III)
Physical Form Crystalline
MDL Number MFCD00058705
CAS 34228-15-4
Synonym Gallium(III)-DPM; Ga(TMHD)3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color White
Melting Point 219° to 220°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.