missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III)

Manufacturer:  Alfa Aesar™ 1449106

 View more versions of this product

Catalog No. AA1449106



Specifications

Specifications

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gallium(III)
White
5g
C33H57GaO6
tris 2,2,6,6-tetramethyl-3,5-heptanedionato gallium iii, gallium iii-dpm, ga tmhd 3, gallium iii 2,2,6,6-tetramethylheptanedionate, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy gallanyl oxy-2,2,6,6-tetramethylhept-4-en-3-one
CC(C)(C)C(=CC(=O)C(C)(C)C)O[Ga](OC(=CC(=O)C(C)(C)C)C(C)(C)C)OC(=CC(=O)C(C)(C)C)C(C)(C)C
619.536
619.54
Hygroscopic
219°C to 220°C
34228-15-4
MFCD00058705
AGZCDNOBXJMLBA-LWTKGLMZSA-K
(Z)-5-bis[[(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl]oxy]gallanyloxy-2,2,6,6-tetramethylhept-4-en-3-one
72376391
Crystalline

Chemical Identifiers

34228-15-4
619.536
AGZCDNOBXJMLBA-LWTKGLMZSA-K
72376391
CC(C)(C)C(=CC(=O)C(C)(C)C)O[Ga](OC(=CC(=O)C(C)(C)C)C(C)(C)C)OC(=CC(=O)C(C)(C)C)C(C)(C)C
C33H57GaO6
MFCD00058705
tris 2,2,6,6-tetramethyl-3,5-heptanedionato gallium iii, gallium iii-dpm, ga tmhd 3, gallium iii 2,2,6,6-tetramethylheptanedionate, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy gallanyl oxy-2,2,6,6-tetramethylhept-4-en-3-one
(Z)-5-bis[[(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl]oxy]gallanyloxy-2,2,6,6-tetramethylhept-4-en-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents