missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gadolinium(III)

Manufacturer:  Alfa Aesar™ 1448606

 View more versions of this product

Catalog No. AA1448606



Specifications

Specifications

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gadolinium(III)
167°C to 170°C
14768-15-1
MFCD00010349
KUKZOIVDRFKQPE-LWTKGLMZSA-N
gadolinium;(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
132984032
Crystalline
Hygroscopic
5g
C33H60GdO6
tris 2,2,6,6-tetramethyl-3,5-heptanedionate gadolinium
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Gd]
710.087
707.07

Chemical Identifiers

14768-15-1
710.087
KUKZOIVDRFKQPE-LWTKGLMZSA-N
132984032
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Gd]
C33H60GdO6
MFCD00010349
tris 2,2,6,6-tetramethyl-3,5-heptanedionate gadolinium
gadolinium;(Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents