missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III)

Manufacturer:  Alfa Aesar™ 8789803

 View more versions of this product

Catalog No. AA8789803



Specifications

Specifications

Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)europium (III)
C33H60EuO6
Europium(III)-DPM; Eu(TMHD)3
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Eu]
704.801
701.78
187°C to 189°C
Hygroscopic
15522-71-1
MFCD00000019
RHXUZKJNHAMZEP-QFVJJVGWSA-N
europium;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
10897800
Crystalline
1g

Chemical Identifiers

15522-71-1
704.801
RHXUZKJNHAMZEP-QFVJJVGWSA-N
10897800
CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Eu]
C33H60EuO6
MFCD00000019
Europium(III)-DPM; Eu(TMHD)3
europium;(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents