Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)erbium(III)

$241.08 - $241.08

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)erbium(III)
CAS 34750-80-6
Formula Weight 717.07
MDL Number MFCD00015705
Synonym Erbium-DPM; Er(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1960203

  SDS
alfa aesar™
1960203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1960206

  SDS
alfa aesar™
1960206
5g Each for $241.08
Description & Specifications

Specifications

Formula Weight 717.07
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)erbium(III)
Physical Form Crystalline
MDL Number MFCD00015705
CAS 34750-80-6
Synonym Erbium-DPM; Er(TMHD)3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 179° to 181°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.