Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III), 98%

Superconductor MOCVD Precursor

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III)
CAS 15522-69-7
Formula Weight 712.32
MDL Number MFCD00000017
Synonym Dysprosium(III)-DPM; Dy(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3646402

  SDS
alfa aesar™
3646402
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3646404

  SDS
alfa aesar™
3646404
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 712.32
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Quantity 0.5g
Melting Point 182° to 183°C
Physical Form Powder
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III)
Synonym Dysprosium(III)-DPM; Dy(TMHD)3
MDL Number MFCD00000017
CAS 15522-69-7
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.