Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99.9% (metals basis)

$406.36 - $406.36

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III)
CAS 14877-41-9
Formula Weight 608.72
MDL Number MFCD00233617
Synonym Cobalt(III)-DPM; Co(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4128006

 
alfa aesar™
4128006
5g Each for $406.36
Description & Specifications

Specifications

Formula Weight 608.72
Assay 99.9%
Synonym Cobalt(III)-DPM; Co(TMHD)3
Sensitivity Hygroscopic
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III)
Physical Form Powder
Assay Percent Notes (metals basis)
MDL Number MFCD00233617
CAS 14877-41-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Color Green
Melting Point 254° to 256°C
Boiling Point 120°C (0.5mmHg) (subl.)
Odor Odorless
Quantity 5g
Health Hazard 3 GHS P Statement
P280h-P262
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
Safety and Handling

Warning

GHS P Statement
P280h-P262
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.