missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%

Vapor deposition

Manufacturer:  Alfa Aesar™ 4399309

 View more versions of this product

Catalog No. AA4399309



Specifications

Specifications

Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt (III)
Green
10g
C33H57CoO6
cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Co+3]
608.746
608.72
99%
120°C (0.5mmHg)(sublimation)
254°C to 256°C
14877-41-9
MFCD00233617
UJRJNARBHKKZST-LWTKGLMZSA-K
cobalt(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
131675252
Powder

Chemical Identifiers

14877-41-9
608.746
UJRJNARBHKKZST-LWTKGLMZSA-K
131675252
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Co+3]
C33H57CoO6
MFCD00233617
cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
cobalt(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents