Learn More
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%, Thermo Scientific Chemicals
Vapor deposition
Supplier: Thermo Scientific Chemicals 04399302
Description
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III) is used precursor for aerosol-assisted CVD of mixed-conducting ceramic films of Sr-Co-Fe perovskite phases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt (III) | |
254°C to 256°C | |
120°C (0.5 mmHg)(sublimation) | |
MFCD00233617,MFCD00233617 | |
cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
UJRJNARBHKKZST-UHFFFAOYSA-K | |
cobalt(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate) | |
131675252 | |
99% |
14877-41-9 | |
Green | |
C33H57CoO6 | |
0.5 g | |
Insoluble in water. | |
[Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C | |
608.75 | |
608.72 | |
Powder |
Chemical Identifiers
14877-41-9 | |
608.75 | |
UJRJNARBHKKZST-UHFFFAOYSA-K | |
131675252 |
C33H57CoO6 | |
MFCD00233617,MFCD00233617 | |
cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
[Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only