Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), 99.9%

$41.55 - $41.55

Chemical Identifiers

Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III)
CAS 142617-53-6
Formula Weight 758.79
MDL Number MFCD00064763
Synonym Bismuth(III)-DPM; Bi(TMHD)3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41202MD

  SDS
alfa aesar™
41202MD
250mg Each for $41.55
Description & Specifications

Specifications

Formula Weight 758.79
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III)
Quantity 250mg
Physical Form Crystalline
Assay 99.9%
MDL Number MFCD00064763
CAS 142617-53-6
Synonym Bismuth(III)-DPM; Bi(TMHD)3
Health Hazard 1 Warning
Boiling Point 150°C (0.05mmHg)
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color White to brown
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 114° to 116°C
Sensitivity Air & Moisture Sensitive
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.