missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(1-pyrazolyl)methane, TCI America™

Manufacturer:  TCI America T30265G

 View more versions of this product

Catalog No. T30265G



Specifications

Specifications

Tris(1-pyrazolyl)methane
104°C
80510-03-8
MFCD00040255
C1=CN(N=C1)C(N2C=CC=N2)N3C=CC=N3
214.232
214.23
≥98.0% (GC)
White-Yellow
5g
C10H10N6
QOHVIMQQEFHOLE-UHFFFAOYSA-N
1-[di(pyrazol-1-yl)methyl]pyrazole
5243808
Crystal-Powder at 20°C

Chemical Identifiers

80510-03-8
214.232
QOHVIMQQEFHOLE-UHFFFAOYSA-N
1-[di(pyrazol-1-yl)methyl]pyrazole
C10H10N6
MFCD00040255
5243808
C1=CN(N=C1)C(N2C=CC=N2)N3C=CC=N3
Safety and Handling

Safety and Handling

TSCA : No

Documents